siRNA Details
| Gene Symbol: FAM24A | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001029888.1 | NP_001025059.1 | 3 | 341 | ||
| XM_017015638.1 | XP_016871127.1 | 3 | 444 | |||
| XM_017015639.1 | XP_016871128.1 | 3 | 349 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

FAM24A| Gene Symbol: FAM24A | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001029888.1 | NP_001025059.1 | 3 | 341 | ||
| XM_017015638.1 | XP_016871127.1 | 3 | 444 | |||
| XM_017015639.1 | XP_016871128.1 | 3 | 349 | |||