siRNA Details
| Gene Symbol: CLRN3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_152311.3 | NP_689524.1 | 3 | 712 | ||
| XM_011539274.2 | XP_011537576.1 | 2 | 531 | |||
| GenBank mRNA | BC029478.1 | - | 3 | 712 | 4733535 | |
| BM830163.1 | - | 3 | 465 | |||
| BX640823.1 | - | 2 | 992 | |||

CLRN3
Genomic Map