siRNA Details
| Gene Symbol: OR5P2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_153444.1 | NP_703145.1 | 1 | 72 | ||
| GenBank mRNA | AF173006.1 | - | 1 | 72 | ||
| BC101710.1 | - | 1 | 103 | 8069216 | ||
| BC113557.1 | - | 1 | 103 | 8322609 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

OR5P2| Gene Symbol: OR5P2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_153444.1 | NP_703145.1 | 1 | 72 | ||
| GenBank mRNA | AF173006.1 | - | 1 | 72 | ||
| BC101710.1 | - | 1 | 103 | 8069216 | ||
| BC113557.1 | - | 1 | 103 | 8322609 | ||