siRNA Details
| Gene Symbol: C12orf54 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_152319.3 | NP_689532.1 | 4, 5 | 264 | ||
| XM_005268636.3 | XP_005268693.1 | 3, 4 | 146 | |||
| XM_011537896.2 | XP_011536198.1 | 6, 7 | 912 | |||
| XM_017018796.1 | XP_016874285.1 | 8, 9 | 1102 | |||
| GenBank mRNA | AY255793.1 | - | 3, 4 | 169 | ||
| AY255794.1 | - | 3, 4 | 169 | |||
| BC031670.2 | - | 3, 4 | 190 | 5165130 | ||
| BQ429029.1 | - | 4, 5 | 267 | |||

C12orf54
Genomic Map