siRNA Details
| Gene Symbol: SLAIN1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001040153.3 | NP_001035243.2 | 4 | 482 | ||
| NM_001242868.1 | NP_001229797.1 | 4 | 991 | |||
| NM_001242871.1 | NP_001229800.1 | 3 | 530 | |||
| NM_144595.3 | NP_653196.1 | 3 | 446 | |||
| XM_011534929.2 | XP_011533231.1 | 5 | 871 | |||
| XM_011534930.1 | XP_011533232.1 | 4 | 384 | |||
| GenBank mRNA | AK054608.1 | - | 3 | 226 | ||
| AK289714.1 | - | 3 | 236 | |||
| AK294201.1 | - | 4 | 482 | |||
| AK295423.1 | - | 3 | 306 | |||
| AL834332.1 | - | 3 | 239 | |||
| BC045177.1 | - | 3 | 255 | |||
| BC111497.1 | - | 3 | 237 | 6148614 | ||
| BG391962.1 | - | 370 | ||||
| BQ433296.1 | - | 5 | 673 | |||
| DA343547.1 | - | 3 | 446 | |||

SLAIN1
Genomic Map