siRNA Details
| Gene Symbol: SOCS4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_080867.2 | NP_543143.1 | 2 | 1081 | ||
| NM_199421.1 | NP_955453.1 | 3 | 1210 | |||
| XM_011536425.1 | XP_011534727.1 | 3 | 1264 | |||
| XM_011536426.1 | XP_011534728.1 | 3 | 1179 | |||
| GenBank mRNA | AF424815.1 | - | 1 | 861 | ||
| AK124083.1 | - | 2 | 934 | |||
| BC060790.1 | - | 3 | 1088 | 30338646 | ||
| BX161418.1 | - | 2 | 977 | |||
| BX648734.1 | - | 2 | 867 | |||
| CR933659.1 | - | 2 | 971 | |||

SOCS4
Genomic Map