siRNA Details
| Gene Symbol: DEGS2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_206918.2 | NP_996801.2 | 2 | 463 | ||
| XM_006720043.3 | XP_006720106.1 | 3 | 478 | |||
| GenBank mRNA | AY541700.1 | - | 2 | 383 | ||
| BC063598.1 | - | 2 | 410 | 4775939 | ||
| BM789564.1 | - | 2 | 463 | |||

DEGS2
Genomic Map