siRNA Details
| Gene Symbol: TMEM219 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001083613.1 | NP_001077082.1 | 4 | 706 | ||
| NM_194280.3 | NP_919256.1 | 4 | 703 | |||
| XM_005255123.1 | XP_005255180.1 | 4 | 705 | |||
| GenBank mRNA | BC017488.1 | - | 3 | 471 | ||
| BC031217.2 | - | 3 | 599 | |||
| BC050051.1 | - | 4 | 733 | 5278133 | ||
| CF453990.1 | - | 650 | ||||
| FJ748884.1 | - | 3 | 566 | |||

TMEM219
Genomic Map