siRNA Details
| Gene Symbol: KRT222 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_152349.2 | NP_689562.1 | 2 | 261 | ||
| GenBank mRNA | AK308598.1 | - | 2 | 325 | ||
| BC032815.1 | - | 2 | 220 | 4346009 | ||
| BX538083.1 | - | 2 | 214 | |||
| DB465511.1 | - | 2 | 261 | |||

KRT222
Genomic Map