siRNA Details
| Gene Symbol: ZSWIM7 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001042697.1 | NP_001036162.1 | 4 | 374 | ||
| NM_001042698.1 | NP_001036163.1 | 4 | 374 | |||
| GenBank mRNA | AK093384.1 | - | 2 | 123 | ||
| BC018807.1 | - | 4 | 345 | |||
| BC044842.1 | - | 4 | 332 | |||
| BC066344.1 | - | 4 | 366 | |||
| BC089386.1 | - | 5 | 458 | |||
| BC094855.1 | - | 6 | 656 | |||
| BU075283.1 | - | 4 | 377 | |||
| DQ530600.1 | - | 4 | 276 | |||

ZSWIM7
Genomic Map