siRNA Details
| Gene Symbol: CLDND2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_152353.2 | NP_689566.1 | 3 | 762 | ||
| XM_011526425.2 | XP_011524727.1 | 3 | 1014 | |||
| XM_011526428.2 | XP_011524730.1 | 3 | 1061 | |||
| XM_017026244.1 | XP_016881733.1 | 2 | 1443 | |||
| XM_017026245.1 | XP_016881734.1 | 3 | 1083 | |||
| XM_017026246.1 | XP_016881735.1 | 2 | 1323 | |||
| XM_017026247.1 | XP_016881736.1 | 2 | 1443 | |||
| GenBank mRNA | BC029518.1 | - | 3 | 762 | 5295481 | |

CLDND2
Genomic Map