siRNA Details
| Gene Symbol: ZNF57 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001319083.1 | NP_001306012.1 | 4 | 737 | ||
| NM_173480.2 | NP_775751.1 | 4 | 836 | |||
| XM_011527682.2 | XP_011525984.1 | 3 | 704 | |||
| GenBank mRNA | AK302165.1 | - | 4 | 737 | ||
| AK314848.1 | - | 4 | 758 | |||
| BC028974.1 | - | 4 | 768 | 5138909 | ||
| BX537601.1 | - | 4 | 777 | |||
| EF534355.1 | - | 4 | 836 | |||

ZNF57
Genomic Map