siRNA Details
| Gene Symbol: LRRC25 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_145256.2 | NP_660299.2 | 1 | 1290 | ||
| XM_005259739.3 | XP_005259796.1 | 2 | 844 | |||
| GenBank mRNA | AJ422148.1 | - | 1 | 977 | ||
| AK095435.1 | - | 1 | 769 | |||
| AY358151.1 | - | 1 | 966 | |||
| BC025744.1 | - | 2 | 803 | |||
| BC053659.1 | - | 2 | 816 | |||
| BC071640.1 | - | 1 | 661 | 5216040 | ||
| BC093842.1 | - | 1 | 702 | 7939687 | ||

LRRC25
Genomic Map