siRNA Details
| Gene Symbol: CCSAP | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_145257.4 | NP_660300.3 | 4 | 849 | ||
| GenBank mRNA | AK291437.1 | - | 3 | 810 | ||
| BC015419.1 | - | 3 | 823 | 4396825 | ||
| BC071609.1 | - | 4 | 895 | 5264741 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

CCSAP| Gene Symbol: CCSAP | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_145257.4 | NP_660300.3 | 4 | 849 | ||
| GenBank mRNA | AK291437.1 | - | 3 | 810 | ||
| BC015419.1 | - | 3 | 823 | 4396825 | ||
| BC071609.1 | - | 4 | 895 | 5264741 | ||