siRNA Details
| Gene Symbol: MYOM3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_152372.3 | NP_689585.3 | 15 | 1882 | ||
| GenBank mRNA | AK125972.1 | - | 13 | 1924 | ||
| AK128233.1 | - | 7 | 942 | |||
| BC067101.1 | - | 15 | 1892 | 5928128 | ||
| BX537911.1 | - | 14 | 1606 | |||
| BX647453.1 | - | 15 | 1843 | |||
| BX647455.1 | - | 15 | 1882 | |||
| BX647644.1 | - | 15 | 1843 | |||

MYOM3
Genomic Map