siRNA Details
| Gene Symbol: ZNF648 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001009992.1 | NP_001009992.1 | 2 | 295 | ||
| XM_017000301.1 | XP_016855790.1 | 4 | 3722 | |||
| XM_017000302.1 | XP_016855791.1 | 6 | 4228 | |||
| GenBank mRNA | AK128654.1 | - | 2 | 295 | ||
| BC137223.1 | - | 2 | 198 | 9021220 | ||
| BC137224.1 | - | 2 | 198 | 9021221 | ||

ZNF648
Genomic Map