siRNA Details
| Gene Symbol: C1orf131 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001300830.1 | NP_001287759.1 | 4 | 623 | ||
| NM_152379.3 | NP_689592.2 | 4 | 626 | |||
| GenBank mRNA | AK303359.1 | - | 4 | 582 | ||
| AL834274.1 | - | 4 | 609 | |||
| BC036800.1 | - | 4 | 603 | 5589128 | ||
| BC053609.1 | - | 4 | 575 | |||
| BC062353.1 | - | 4 | 606 | 4648754 | ||

C1orf131
Genomic Map