siRNA Details
| Gene Symbol: C1orf131 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001300830.1 | NP_001287759.1 | 7 | 877 | ||
| NM_152379.3 | NP_689592.2 | 7 | 880 | |||
| GenBank mRNA | AL834274.1 | - | 6 | 1919 | ||
| AY358235.1 | - | 1 | 830 | |||
| BC036800.1 | - | 7 | 857 | 5589128 | ||
| BC053609.1 | - | 7 | 829 | |||
| BC062353.1 | - | 7 | 860 | 4648754 | ||

C1orf131
Genomic Map