siRNA Details
| Gene Symbol: SIRPD | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_178460.2 | NP_848555.2 | 1 | 95 | ||
| GenBank mRNA | AK093083.1 | - | 1 | 1241 | ||
| BC033502.1 | - | 1 | 95 | 5172531 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

SIRPD| Gene Symbol: SIRPD | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_178460.2 | NP_848555.2 | 1 | 95 | ||
| GenBank mRNA | AK093083.1 | - | 1 | 1241 | ||
| BC033502.1 | - | 1 | 95 | 5172531 | ||