siRNA Details
| Gene Symbol: MIR1-1HG | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001302812.1 | NP_001289741.1 | 5 | 633 | ||
| NM_178463.3 | NP_848558.1 | 4 | 1014 | |||
| GenBank mRNA | AK056985.1 | - | 4 | 1030 | ||
| BC110595.2 | - | 4 | 689 | 40022808 | ||

MIR1-1HG
Genomic Map