siRNA Details
Gene Symbol: PIGU | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_080476.4 | NP_536724.1 | 11 | 1346 | ||
XM_011528542.2 | XP_011526844.1 | 5 | 637 | |||
XM_017027664.1 | XP_016883153.1 | 10 | 1020 | |||
GenBank mRNA | AB086842.1 | - | 11 | 1152 | ||
AK075507.1 | - | 11 | 1171 | |||
AK301900.1 | - | 11 | 1146 | |||
AY339061.1 | - | 10 | 1086 | |||
AY358816.1 | - | 11 | 1176 | |||
AY422169.1 | - | 11 | 1346 | |||
BC030512.1 | - | 11 | 1157 | 4942673 |
