siRNA Details
| Gene Symbol: PIGU | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_080476.4 | NP_536724.1 | 6 | 700 | ||
| XM_017027664.1 | XP_016883153.1 | 6 | 518 | |||
| XM_017027665.1 | XP_016883154.1 | 6 | 518 | |||
| GenBank mRNA | AB086842.1 | - | 6 | 506 | ||
| AK075507.1 | - | 6 | 525 | |||
| AK301900.1 | - | 6 | 500 | |||
| AK308627.1 | - | 5 | 447 | |||
| AY339061.1 | - | 5 | 440 | |||
| AY358816.1 | - | 6 | 530 | |||
| AY422169.1 | - | 6 | 700 | |||
| BC030512.1 | - | 6 | 511 | 4942673 | ||
More Information
Other Species Targeted
This siRNA also targets the following transcripts in Mouse: NM_001004721, BC120682, CK334159, AK211799, BC120680, and others.

PIGU
Genomic Map