siRNA Details
| Gene Symbol: ZNF280A | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_080740.4 | NP_542778.2 | 2 | 407 | ||
| GenBank mRNA | AK093108.1 | - | 2 | 401 | ||
| BC053901.1 | - | 2 | 381 | 6166804 | ||
| BP283207.1 | - | 2 | 406 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

ZNF280A| Gene Symbol: ZNF280A | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_080740.4 | NP_542778.2 | 2 | 407 | ||
| GenBank mRNA | AK093108.1 | - | 2 | 401 | ||
| BC053901.1 | - | 2 | 381 | 6166804 | ||
| BP283207.1 | - | 2 | 406 | |||