siRNA Details
| Gene Symbol: TRABD2A | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001080824.2 | NP_001074293.1 | 2 | 506 | ||
| NM_001277053.1 | NP_001263982.1 | 2 | 506 | |||
| NM_001307978.1 | NP_001294907.1 | 2 | 506 | |||
| XM_011532504.1 | XP_011530806.1 | 2 | 306 | |||
| GenBank mRNA | AK296608.1 | - | 2 | 343 | ||
| AK309615.1 | - | 2 | 345 | |||
| BC051789.1 | - | 2 | 507 | |||
| JQ653415.1 | - | 2 | 300 | |||

TRABD2A
Genomic Map