siRNA Details
| Gene Symbol: CIB4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001029881.2 | NP_001025052.1 | 3, 4 | 239 | ||
| XM_011532514.2 | XP_011530816.1 | 3, 4 | 322 | |||
| XM_017003329.1 | XP_016858818.1 | 2, 3 | 1093 | |||
| GenBank mRNA | BC127684.1 | - | 3, 4 | 239 | 40127392 | |
| BC127685.1 | - | 3, 4 | 237 | 40127393 | ||
| BC140923.1 | - | 3, 4 | 213 | 9056735 | ||

CIB4
Genomic Map