siRNA Details
| Gene Symbol: C2orf50 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_182500.2 | NP_872306.1 | 3 | 601 | ||
| XM_006711857.2 | XP_006711920.1 | 3 | 525 | |||
| XM_006711858.2 | XP_006711921.1 | 3 | 525 | |||
| XM_006711859.3 | XP_006711922.1 | 3 | 525 | |||
| GenBank mRNA | AK057872.1 | - | 3 | 607 | ||
| AK292828.1 | - | 3 | 526 | |||
| BC120930.2 | - | 3 | 568 | 40114857 | ||

C2orf50
Genomic Map