siRNA Details
| Gene Symbol: ADH6 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_000672.3 | NP_000663.1 | 5 | 600 | ||
| NM_001102470.1 | NP_001095940.1 | 5 | 600 | |||
| GenBank mRNA | AK092768.1 | - | 5 | 600 | ||
| AK298290.1 | - | 1 | 169 | |||
| BC039065.1 | - | 5 | 589 | 5184104 | ||

ADH6
Genomic Map