siRNA Details
| Gene Symbol: CRYBG3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_153605.3 | NP_705833.3 | 11 | 7653 | ||
| XM_005247117.4 | XP_005247174.1 | 10 | 7356 | |||
| GenBank mRNA | AK091199.1 | - | 3 | 124 | ||
| BX647079.1 | - | 3 | 141 | |||
| CR749313.1 | - | 8 | 778 | |||

CRYBG3
Genomic Map