siRNA Details
| Gene Symbol: ERICH6 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001308234.1 | NP_001295163.1 | 13 | 1409 | ||
| NM_152394.4 | NP_689607.2 | 13 | 1713 | |||
| GenBank mRNA | AK093375.1 | - | 14 | 1548 | ||
| AL834214.1 | - | 1213 | ||||
| BC029577.1 | - | 13 | 1393 | 5268183 | ||
| BC036239.1 | - | 13 | 1697 | 5297417 | ||

ERICH6
Genomic Map