siRNA Details
| Gene Symbol: PPM1M | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001122870.2 | NP_001116342.1 | 6, 7 | 639 | ||
| NM_144641.3 | NP_653242.3 | 7, 8 | 1005 | |||
| XM_005264879.2 | XP_005264936.1 | 7, 8 | 1073 | |||
| XM_005264880.3 | XP_005264937.1 | 6, 7 | 818 | |||
| XM_005264881.1 | XP_005264938.1 | 7, 8 | 894 | |||
| GenBank mRNA | AB474372.1 | - | 7, 8 | 973 | ||
| AK056894.1 | - | 7, 8 | 894 | |||
| AK129647.1 | - | 1, 2 | 270 | |||
| AL832484.1 | - | 5, 6 | 913 | |||
| BC009644.1 | - | 6, 7 | 600 | 3905720 | ||

PPM1M
Genomic Map