siRNA Details
| Gene Symbol: COX6A2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_005205.3 | NP_005196.1 | 3 | 342 | ||
| GenBank mRNA | BC029818.1 | - | 3 | 302 | 5174639 | |
| BP319904.1 | - | 342 | ||||
| F27891.1 | - | 3 | 305 | |||
| HE647858.1 | - | 3 | 239 | |||
| M83308.1 | - | 3 | 275 | |||

COX6A2
Genomic Map