siRNA Details
| Gene Symbol: SVOPL | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001139456.1 | NP_001132928.1 | 13, 14 | 1360 | ||
| NM_174959.3 | NP_777619.1 | 9, 10 | 977 | |||
| XM_005250143.3 | XP_005250200.1 | 14, 15 | 1764 | |||
| XM_011515797.2 | XP_011514099.1 | 10, 11 | 1051 | |||
| XM_017011746.1 | XP_016867235.1 | 11, 12 | 1166 | |||
| XM_017011747.1 | XP_016867236.1 | 10, 11 | 991 | |||
| XM_017011748.1 | XP_016867237.1 | 10, 11 | 996 | |||
| GenBank mRNA | BC036796.1 | - | 9, 10 | 977 | 5590416 | |
| BX349158.2 | - | 4, 5 | 407 | |||

SVOPL
Genomic Map