siRNA Details
| Gene Symbol: ASZ1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001301821.1 | NP_001288750.1 | 9, 10 | 997 | ||
| NM_001301822.1 | NP_001288751.1 | 8, 9 | 885 | |||
| NM_130768.2 | NP_570124.1 | 9, 10 | 997 | |||
| GenBank mRNA | AF461259.1 | - | 9, 10 | 963 | ||
| AK093445.1 | - | 9, 10 | 950 | |||
| BC028695.1 | - | 9, 10 | 967 | |||
| BC034963.1 | - | 9, 10 | 972 | 4824818 | ||
| BC068488.1 | - | 8, 9 | 858 | |||
| BC126186.1 | - | 9, 10 | 956 | 8991902 | ||
| BC126188.1 | - | 9, 10 | 956 | 8991904 | ||
| BC144212.1 | - | 9, 10 | 956 | 9052732 | ||

ASZ1
Genomic Map