siRNA Details
| Gene Symbol: CR2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001006658.2 | NP_001006659.1 | 8, 9 | 1622 | ||
| NM_001877.4 | NP_001868.2 | 8, 9 | 1622 | |||
| XM_011509206.2 | XP_011507508.1 | 8, 9 | 1518 | |||
| GenBank mRNA | AK223627.1 | - | 8, 9 | 1594 | ||
| AK301496.1 | - | 8, 9 | 1590 | |||
| BC090937.1 | - | 8, 9 | 1557 | 3066233 | ||
| BC136394.1 | - | 8, 9 | 1596 | 9020381 | ||
| J03565.1 | - | 8, 9 | 1566 | |||
| M26004.1 | - | 8, 9 | 1572 | |||
| Y00649.1 | - | 8, 9 | 1555 | |||

CR2
Genomic Map