siRNA Details
| Gene Symbol: C9orf57 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001128618.1 | NP_001122090.1 | 2 | 194 | ||
| GenBank mRNA | BC036255.1 | - | 2 | 194 | ||
| BC044620.1 | - | 2 | 172 | |||
| BC053956.1 | - | 2 | 172 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

C9orf57| Gene Symbol: C9orf57 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001128618.1 | NP_001122090.1 | 2 | 194 | ||
| GenBank mRNA | BC036255.1 | - | 2 | 194 | ||
| BC044620.1 | - | 2 | 172 | |||
| BC053956.1 | - | 2 | 172 | |||