siRNA Details
| Gene Symbol: PIP5KL1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001135219.1 | NP_001128691.1 | 9 | 897 | ||
| NM_173492.1 | NP_775763.1 | 4 | 453 | |||
| XM_017014279.1 | XP_016869768.1 | 8 | 1945 | |||
| GenBank mRNA | BC042184.1 | - | 4 | 453 | 5163193 | |

PIP5KL1
Genomic Map