siRNA Details
| Gene Symbol: MAGEE2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_138703.4 | NP_619648.1 | 1 | 1554 | ||
| GenBank mRNA | AF490509.1 | - | 1 | 1536 | ||
| AK094624.1 | - | 1 | 1546 | |||
| BC093964.1 | - | 1 | 1455 | 7939809 | ||
| BC111949.1 | - | 1 | 1455 | 8327417 | ||

MAGEE2
Genomic Map