siRNA Details
| Gene Symbol: BRI3BP | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_080626.5 | NP_542193.3 | 3 | 604 | ||
| GenBank mRNA | AF283671.2 | - | 3 | 343 | ||
| AF284094.1 | - | 3 | 548 | |||
| AF441865.1 | - | 2 | 209 | |||
| AK055703.1 | - | 1 | 125 | |||
| BC034525.1 | - | 3 | 581 | 4778946 | ||

BRI3BP
Genomic Map