siRNA Details
| Gene Symbol: SOGA1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_080627.3 | NP_542194.2 | 7 | 2892 | ||
| NM_199181.2 | NP_954650.2 | 7 | 2165 | |||
| GenBank mRNA | AK126630.1 | - | 7 | 2165 | ||
| AK131491.1 | - | 3 | 1392 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

SOGA1| Gene Symbol: SOGA1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_080627.3 | NP_542194.2 | 7 | 2892 | ||
| NM_199181.2 | NP_954650.2 | 7 | 2165 | |||
| GenBank mRNA | AK126630.1 | - | 7 | 2165 | ||
| AK131491.1 | - | 3 | 1392 | |||