siRNA Details
| Gene Symbol: TLDC2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_080628.2 | NP_542195.1 | 3, 4 | 401 | ||
| XM_017027672.1 | XP_016883161.1 | 3, 4 | 391 | |||
| XM_017027674.1 | XP_016883163.1 | 2, 3 | 241 | |||
| GenBank mRNA | AK123544.1 | - | 3, 4 | 355 | ||
| BC127688.1 | - | 3, 4 | 401 | 40127440 | ||
| BC130646.1 | - | 3, 4 | 390 | 40146677 | ||
| BC130648.1 | - | 3, 4 | 390 | 40146679 | ||

TLDC2
Genomic Map