siRNA Details
Gene Symbol: CSF2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_000758.3 | NP_000749.2 | 4 | 386 | ||
GenBank mRNA | BC108724.1 | - | 4 | 373 | 3886571 | |
BC113999.1 | - | 4 | 377 | 40084798 | ||
KP271009.1 | - | 4 | 303 | |||
M11220.1 | - | 4 | 386 | |||
M11734.1 | - | 4 | 369 |
