siRNA Details
| Gene Symbol: WDR66 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001178003.1 | NP_001171474.1 | 4 | 949 | ||
| NM_144668.5 | NP_653269.3 | 4 | 936 | |||
| GenBank mRNA | AK097102.1 | - | 3 | 690 | ||
| AL833930.1 | - | 2 | 224 | |||
| BC028421.2 | - | 4 | 934 | 4826893 | ||
| BC036233.1 | - | 4 | 929 | 5295406 | ||

WDR66
Genomic Map