siRNA Details
| Gene Symbol: TSPAN19 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001100917.1 | NP_001094387.1 | 6 | 483 | ||
| XM_005268669.4 | XP_005268726.1 | 6 | 533 | |||
| XM_017018862.1 | XP_016874351.1 | 5 | 421 | |||
| XM_017018863.1 | XP_016874352.1 | 5 | 421 | |||
| XM_017018864.1 | XP_016874353.1 | 5 | 421 | |||
| GenBank mRNA | AK126325.1 | - | 6 | 479 | ||
| CB852874.1 | - | 4 | 549 | |||
| CB854430.1 | - | 4 | 543 | |||

TSPAN19
Genomic Map