siRNA Details
| Gene Symbol: CSNK2A1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001895.3 | NP_001886.1 | 10 | 1076 | ||
| NM_177559.2 | NP_808227.1 | 11 | 1193 | |||
| NM_177560.2 | NP_808228.1 | 9 | 866 | |||
| GenBank mRNA | AB451279.1 | - | 9 | 800 | ||
| AB451411.1 | - | 9 | 800 | |||
| AK302583.1 | - | 9 | 755 | |||
| AK312535.1 | - | 11 | 1084 | |||
| BC011668.2 | - | 10 | 948 | 4121882 | ||
| BC050036.1 | - | 10 | 967 | 5298655 | ||
| BC053532.1 | - | 9 | 895 | 3908058 | ||
| BC071167.1 | - | 9 | 807 | 5772615 | ||
| BT019792.1 | - | 9 | 800 | |||
| DQ354583.1 | - | 10 | 942 | |||
| J02853.1 | - | 10 | 953 | |||
| M55265.1 | - | 10 | 948 | |||
| S53149.1 | - | 10 | 953 | |||
| Gene Symbol: CSNK2A3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001256686.1 | NP_001243615.1 | 1 | 1038 | ||

CSNK2A1
Genomic Map