siRNA Details
| Gene Symbol: OTOA | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001161683.1 | NP_001155155.1 | 12 | 1646 | ||
| NM_144672.3 | NP_653273.3 | 16 | 1790 | |||
| NM_170664.2 | NP_733764.1 | 7 | 884 | |||
| XM_011545747.2 | XP_011544049.1 | 17 | 2169 | |||
| XM_011545748.2 | XP_011544050.2 | 6 | 1163 | |||
| XM_017022950.1 | XP_016878439.1 | 17 | 1893 | |||
| XM_017022951.1 | XP_016878440.1 | 2 | 187 | |||
| GenBank mRNA | AK093062.1 | - | 7 | 884 | ||
| AK125840.1 | - | 12 | 1646 | |||
| AK302733.1 | - | 6 | 946 | |||
| BC129992.1 | - | 16 | 1787 | |||
| BC129993.1 | - | 16 | 1794 | 40128985 | ||

OTOA
Genomic Map