siRNA Details
| Gene Symbol: CARMIL2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001013838.1 | NP_001013860.1 | 2 | 195 | ||
| NM_001317026.1 | NP_001303955.1 | 2 | 195 | |||
| XM_011522874.1 | XP_011521176.1 | 2 | 318 | |||
| XM_011522875.2 | XP_011521177.2 | 2 | 318 | |||
| XM_017022953.1 | XP_016878442.1 | 2 | 318 | |||
| GenBank mRNA | AB113647.1 | - | 2 | 195 | ||
| AL832446.1 | - | 1 | 47 | |||
| FJ026014.1 | - | 2 | 75 | |||

CARMIL2
Genomic Map