siRNA Details
| Gene Symbol: PRSS36 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001258290.1 | NP_001245219.1 | 13 | 2181 | ||
| NM_173502.4 | NP_775773.2 | 13 | 2196 | |||
| XM_011545758.2 | XP_011544060.1 | 12 | 2414 | |||
| XM_017022968.1 | XP_016878457.1 | 13 | 2185 | |||
| GenBank mRNA | AJ627034.1 | - | 13 | 2168 | ||
| AK075142.1 | - | 13 | 2169 | |||
| AK290310.1 | - | 13 | 2196 | |||
| AK297997.1 | - | 11 | 2137 | |||
| BC137396.1 | - | 13 | 2163 | 9021394 | ||
| BC144615.1 | - | 13 | 2148 | 9053146 | ||

PRSS36
Genomic Map