siRNA Details
| Gene Symbol: CDRT15 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001007530.1 | NP_001007531.1 | 1 | 111 | ||
| GenBank mRNA | BC146859.1 | - | 1 | 137 | 9056787 | |
| Gene Symbol: CDRT15P1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_003261.1 | 1 | 127 | |||
| GenBank mRNA | AA453375.1 | - | 1 | 129 | ||
| BX118788.1 | - | 1 | 128 | |||

CDRT15
Genomic Map