siRNA Details
| Gene Symbol: CDRT15 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001007530.1 | NP_001007531.1 | 3 | 451 | ||
| GenBank mRNA | BC146859.1 | - | 3 | 477 | 9056787 | |
Please enter the information below and press OK to send your cart to Core Services for purchase.

CDRT15| Gene Symbol: CDRT15 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001007530.1 | NP_001007531.1 | 3 | 451 | ||
| GenBank mRNA | BC146859.1 | - | 3 | 477 | 9056787 | |