siRNA Details
| Gene Symbol: C19orf84 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001193623.1 | NP_001180552.1 | 2 | 293 | ||
| NR_126530.1 | 2 | 422 | ||||
| GenBank mRNA | BC043532.1 | - | 2 | 293 | ||
| BC085012.1 | - | 2 | 266 | |||
| BC143997.1 | - | 2 | 411 | |||

C19orf84
Genomic Map